missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methyl-1-benzothiophene, 97%, Thermo Scientific™

Product Code. 10799643
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10799643 1 g 1g
10536314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10799643 Supplier Thermo Scientific Maybridge Supplier No. MO07070DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 111860-00-5
Molecular Formula C9H7BrS
Molecular Weight (g/mol) 227.119
MDL Number MFCD05865140
InChI Key HZPBPLLBZYSGBI-UHFFFAOYSA-N
Synonym 2-bromo-5-methylbenzo b thiophene, benzo b thiophene,2-bromo-5-methyl, acmc-20mexj, 2-bromanyl-5-methyl-1-benzothiophene, 2-bromo-5-methyl-1-benzo b thiophene, benzo b thiophene, 2-bromo-5-methyl
PubChem CID 2794769
IUPAC Name 2-bromo-5-methyl-1-benzothiophene
SMILES CC1=CC2=C(C=C1)SC(=C2)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 227.12
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methyl-1-benzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.