missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylpyridine, 98+%

Product Code. 11466087
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466087 100 g 100g
11446087 5 g 5g
11456087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Codice prodotto. 11466087 Fornitore Thermo Scientific Alfa Aesar N. del fornitore B25083.22
 Restricted Item
Estimated Shipment: 22-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 3510-66-5 | C6H6BrN | 172.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3510-66-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00209553
InChI Key YWNJQQNBJQUKME-UHFFFAOYSA-N
Synonym 2-bromo-5-picoline, 6-bromo-3-picoline, 2-bromo-5-methyl-pyridine, pyridine, 2-bromo-5-methyl, 2-bromo-5-methyl pyridine, 5-methyl-2-bromopyridine, 2-brom-5-methylpyridin, pubchem1185
PubChem CID 564216
IUPAC Name 2-bromo-5-methylpyridine
SMILES CC1=CN=C(C=C1)Br

Specifica

Melting Point 40°C to 45°C
Boiling Point 95°C to 96°C (12 mmHg)
Flash Point 103°C (217°F)
Quantity 100 g
Beilstein 107323
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-methylpyridine

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.