missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylthiazole, 98%

Product Code. 15440949
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440949 5 g 5g
15430949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440949 Supplier Thermo Scientific Alfa Aesar Supplier No. H63951.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 41731-23-1 | C4H4BrNS | 178.047 g/mol

2-Bromo-5-methylthiazole is a brominated thiazole compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-methylthiazole is a brominated thiazole compound for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41731-23-1
Molecular Formula C4H4BrNS
Molecular Weight (g/mol) 178.047
MDL Number MFCD08460610
InChI Key FJPZHYAYNAUKKA-UHFFFAOYSA-N
Synonym 2-bromo-5-methylthiazole, 2-bromo-5-methyl-thiazole, thiazole, 2-bromo-5-methyl, pubchem20436, acmc-209jlg, 2-bromo-5-methyl thiazole, ksc493s9j
PubChem CID 21906106
IUPAC Name 2-bromo-5-methyl-1,3-thiazole
SMILES CC1=CN=C(S1)Br

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 178.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-methylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.