missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiazole, 98%

Product Code. 11437716
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437716 5 g 5g
11427716 1 g 1g
11447716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437716 Supplier Thermo Scientific Alfa Aesar Supplier No. A14701.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3034-48-8 | C3HBrN2O2S | 209.02 g/mol

2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-48-8
Molecular Formula C3HBrN2O2S
Molecular Weight (g/mol) 209.02
MDL Number MFCD00005317
InChI Key ANIJFZVZXZQFDH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrothiazole, thiazole, 2-bromo-5-nitro, 5-nitro-2-bromothiazole, pubchem11070, acmc-1ad6f, 2-bromo-5-nitro-thiazole, 4-27-00-00963 beilstein handbook reference, 2-bromo-5-nitrothiozole, #
PubChem CID 18211
IUPAC Name 2-bromo-5-nitro-1,3-thiazole
SMILES [O-][N+](=O)C1=CN=C(Br)S1

Specifications

Melting Point 85°C to 90°C
Quantity 5 g
Beilstein 4856
Solubility Information Slightly soluble in water.
Formula Weight 209.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitrothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.