missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-(trifluoromethyl)pyridine, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-5-(trifluoromethyl)pyridine |
| CAS | 50488-42-1 |
| Molecular Formula | C6H3BrF3N |
| Quantity | 50 g |
| Synonym | 5-(Trifluoromethyl)-2-bromopyridine |
| InChI Formula | InChI=1S/C6H3BrF3N/c7-5-2-1-4(3-11-5)6(8,9)10/h1-3H |
| SMILES | OC(=O)C(C)(C)NC(=O)[C@@H]1[C@]2(C)[C@H](CC1)[C@H]3[C@@H](CC2)[C@]4(C)[C@@H](NC(=O)C=C4)CC3 |
| IUPAC Name | 2-bromo-5-(trifluoromethyl)pyridine |
| Molecular Weight (g/mol) | 225.99 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction