missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(1H-pyrazol-1-yl)pyridine, ≥97%, Thermo Scientific™

Product Code. 10197762
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10197762 250 mg Amber Glass Bottle 250mg
10738543 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197762 Supplier Thermo Scientific Maybridge Supplier No. CC35110CB
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 123640-41-5
Molecular Formula C8H6BrN3
Molecular Weight (g/mol) 224.061
MDL Number MFCD07368535
InChI Key LPRJOZJWALGJGM-UHFFFAOYSA-N
PubChem CID 7060529
IUPAC Name 2-bromo-6-pyrazol-1-ylpyridine
SMILES C1=CC(=NC(=C1)Br)N2C=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-bromo-6-1h-pyrazol-1-yl pyridine, 2-bromo-6-pyrazol-1-yl pyridine, 2-bromo-6-pyrazol-1-yl-pyridine, pyridine,2-bromo-6-1h-pyrazol-1-yl, pyridine, 2-bromo-6-1h-pyrazol-1-yl, acmc-209apf, 6-bromo-2-pyridyl pyrazole, 2-bromo-6-1-pyrazolyl pyridine, 2-bromanyl-6-pyrazol-1-yl-pyridine, 1-6-bromo-2-pyridinyl-1h-pyrazole
Formula Weight 224.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-(1H-pyrazol-1-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.