missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-fluoroaniline, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-6-fluoroaniline |
| CAS | 65896-11-9 |
| Molecular Formula | C6H5BrFN |
| Quantity | 5 g |
| Synonym | 2-Bromo-6-fluorobenzenamine |
| InChI Formula | InChI=1S/C6H5BrFN/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2 |
| SMILES | Nc1c(F)cccc1Br |
| IUPAC Name | 2-bromo-6-fluoroaniline |
| Molecular Weight (g/mol) | 190.01 |
| Formula Weight | 188.9589 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction