missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(hydroxymethyl)pyridine, 96%

Product Code. 10234884 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10234884

Brand: Thermo Scientific Acros 426310050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 33674-96-3 | C6H6BrNO | 188.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33674-96-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04039884
InChI Key XDDGKNRSCDEWBR-UHFFFAOYSA-N
Synonym 6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
PubChem CID 3517811
IUPAC Name (6-bromopyridin-2-yl)methanol
SMILES OCC1=CC=CC(Br)=N1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 34°C to 39°C
Color White
Boiling Point 246°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 188.02
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 2-Bromo-6-(hydroxymethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-6-(hydroxymethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.