missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxynaphthalene, 98%

Product Code. 11414037
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414037 50 g 50g
11404037 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414037 Supplier Thermo Scientific Alfa Aesar Supplier No. A19450.18
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5111-65-9 | C11H9BrO | 237.096 g/mol

2-Bromo-6-methoxynaphthalene is used in the synthesis of nabumetone [4-(6-methoxy-2-naphthalenyl)-2-butanone] by Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-methoxynaphthalene is used in the synthesis of nabumetone [4-(6-methoxy-2-naphthalenyl)-2-butanone] by Heck reaction.

Solubility
Soluble in DMSO. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5111-65-9
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.096
MDL Number MFCD00004062
InChI Key AYFJBMBVXWNYLT-UHFFFAOYSA-N
Synonym 6-methoxy-2-bromonaphthalene, naphthalene, 2-bromo-6-methoxy, 6-bromo-2-methoxynaphthalene, 2-bromo-6-methoxy-naphthalene, 6-brom-2-naphthyl-methyl ether, 2-bromo-6-methoxynapthalene, unii-s19t080mrg, naproxen ep impurity n, bromo 2--6-methoxy naphthalene, 2-methoxy-6-bromonaphthalene
PubChem CID 78786
IUPAC Name 2-bromo-6-methoxynaphthalene
SMILES COC1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 105°C to 110°C
Boiling Point 160°C to 164°C (3 mmHg)
Quantity 50 g
Beilstein 2043874
Solubility Information Soluble in DMSO. Insoluble in water.
Formula Weight 237.1
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-methoxynaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.