missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-p-xylene, 97%

Product Code. 10644852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10644852 100 g 100g
10449470 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10644852 Supplier Thermo Scientific Acros Supplier No. 173421000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 553-94-6 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-94-6
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000074
InChI Key QXISTPDUYKNPLU-UHFFFAOYSA-N
Synonym 2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
PubChem CID 11121
IUPAC Name 2-bromo-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(Br)=C1

Specifications

Melting Point 9°C to 10°C
CAS Min % 96.0
Color Colorless
CAS Max % 100.0
Density 1.3400g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5495 to 1.5515
Assay Percent Range 96% min. (GC)
Specific Gravity 1.34
Linear Formula (CH3)2C6H3Br
Beilstein 05, 385
Boiling Point 199°C to 201°C
Formula Weight 185.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-p-xylene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-p-xylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.