missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetamide, 98%

Product Code. 10551511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10551511 25 g Glass bottle 25g
10221143 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10551511 Supplier Thermo Scientific Acros Supplier No. 291100250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 683-57-8 | C2H4BrNO | 137.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-57-8
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.96
MDL Number MFCD00008025
InChI Key JUIKUQOUMZUFQT-UHFFFAOYSA-N
Synonym bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
PubChem CID 69632
IUPAC Name 2-bromoacetamide
SMILES C(C(=O)N)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CONH2
Quantity 25 g
Beilstein 02, 216
Formula Weight 137.96
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoacetamide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromoacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.