missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetic Acid Phenylmethyl Ester, TRC

Product Code. 30343595
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30343595

Brand: Toronto Research Chemicals TRCB67911510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoacetic Acid Phenylmethyl Ester
CAS 5437-45-6
Molecular Formula C9H9BrO2
Quantity 10 g
Synonym Bromoacetic Acid Benzyl Ester, 2-Bromoacetic Acid Benzyl Ester, Benzyl 2-Bromoacetate, Benzyl Bromoacetate, Bromoacetic Acid Benzyl Ester, Merbac 35, NSC 16114, NSC 23980, Phenylmethyl Bromoacetate
InChI Formula InChI=1S/C9H9BrO2/c10-6-9(11)12-7-8-4-2-1-3-5-8/h1-5H,6-7H2
SMILES BrCC(=O)OCc1ccccc1
IUPAC Name benzyl 2-bromoacetate
Molecular Weight (g/mol) 229.07
Formula Weight 227.9786
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.