missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetophenone, 98%

Product Code. 11433057
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433057 500 g 500g
11413057 25 g 25g
11423057 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433057 Supplier Thermo Scientific Alfa Aesar Supplier No. A15576.36
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 70-11-1 | C8H7BrO | 199.047 g/mol

It is used in preparation of crystalline esters from acids. 2-Bromoacetophenone is a complete and irreversible inactivator of human liver aldehyde dehydrogenase isoenzymes E1 and E2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of crystalline esters from acids. 2-Bromoacetophenone is a complete and irreversible inactivator of human liver aldehyde dehydrogenase isoenzymes E1 and E2.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids & bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-11-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD00000195
InChI Key LIGACIXOYTUXAW-UHFFFAOYSA-N
Synonym 2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
PubChem CID 6259
ChEBI CHEBI:51846
IUPAC Name 2-bromo-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 48°C to 51°C
Density 1.65
Boiling Point 140°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 500 g
UN Number UN2645
Beilstein 606474
Merck Index 14,1402
Solubility Information Insoluble in water.
Formula Weight 199.05
Percent Purity 98%
Chemical Name or Material 2-Bromoacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.