missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetophenone, 98+%

Product Code. 11984871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11984871 100 g Glass bottle 100g
10622413 5 g Glass bottle 5g
10772221 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11984871 Supplier Thermo Scientific Acros Supplier No. 179761000
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2142-69-0 | C8H7BrO | 199.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-69-0
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000067
InChI Key PIMNFNXBTGPCIL-UHFFFAOYSA-N
Synonym 2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
PubChem CID 75060
IUPAC Name 1-(2-bromophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1Br

Specifications

Color Orange to Yellow
Density 1.4760g/mL
Boiling Point 116.0°C to 117.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH3
Refractive Index 1.5650 to 1.5690
Quantity 100 g
Specific Gravity 1.476
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chlorofom,eth
Formula Weight 199.05
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2'-Bromoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Bromoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.