missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromoacetyl)thiophene, 97%, Thermo Scientific Chemicals

Product Code. 15497937
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497937 5 g 5g
15487937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497937 Supplier Thermo Scientific Alfa Aesar Supplier No. H51060.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10531-41-6 | Molecular Formula: C6H5BrOS | Molecular Weight: 205.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 10531-41-6
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.069
MDL Number MFCD02677721
InChI Key UHWNENCHFSDZQP-UHFFFAOYSA-N
Synonym 2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
PubChem CID 2776372
IUPAC Name 2-bromo-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)CBr

Spezifikation

Melting Point 29°C to 33°C
Density 1.62
Boiling Point 127°C to 130°C (7 mmHg)
Odor Pungent
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Formula Weight 205.07
Percent Purity 97%
Chemical Name or Material 2-(Bromoacetyl)thiophene

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.