missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacrylic acid, 95%

Product Code. 11324504
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324504 1 g 1g
11334504 5 g 5g
15488789 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324504 Supplier Thermo Scientific Alfa Aesar Supplier No. L01194.03
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10443-65-9 | C3H3BrO2 | 150.96 g/mol

2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

Solubility
It is miscible with water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10443-65-9
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.96
MDL Number MFCD00014333
InChI Key HMENQNSSJFLQOP-UHFFFAOYSA-N
Synonym 2-bromoacrylic acid, bromoacrylic acid, ccris 5478, .alpha.-bromoacrylic acid, 2-bromo-acrylic acid, alpha-bromo acrylic acid, 2-propenoic acid, bromo, acmc-1c0k4, 2-propenoic acid, 2-bromo
PubChem CID 82633
IUPAC Name 2-bromoprop-2-enoic acid
SMILES OC(=O)C(Br)=C

Specifications

Melting Point 65°C to 70°C
Quantity 1 g
UN Number UN3261
Beilstein 1740582
Formula Weight 150.96
Percent Purity 95%
Chemical Name or Material 2-Bromoacrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.