missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzo[b]thiophene, ≥97%, Thermo Scientific™

Product Code. 10728815
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10728815 250 mg 250mg
10633615 1 g 1g
10026704 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10728815 Supplier Thermo Scientific Maybridge Supplier No. CC12210CB
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5394-13-8
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD08435846
InChI Key WIFMYMXKTAVDSQ-UHFFFAOYSA-N
Synonym 2-bromobenzo b thiophene, benzo b thiophene, 2-bromo, 2-bromobenzothiophene, bromobenzothiophene, 2-bromobenzthiophene, bromobenzo b thiophene, 2-bromo-benzo b thiophene, 2-bromobenzo b-thiophene
PubChem CID 94786
IUPAC Name 2-bromo-1-benzothiophene
SMILES C1=CC=C2C(=C1)C=C(S2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 213.1
Percent Purity 97%
Chemical Name or Material 2-Bromobenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.