missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoyl Chloride, TRC

Product Code. 30335187
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335187 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335187 Supplier Toronto Research Chemicals Supplier No. TRCB68038025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromobenzoyl Chloride
CAS 7154-66-7
Molecular Formula C7H4BrClO
Quantity 25 g
Synonym o-Bromobenzoyl Chloride, 2-Bromobenzoic Acid Chloride, NSC 73706
InChI Formula InChI=1S/C7H4BrClO/c8-6-4-2-1-3-5(6)7(9)10/h1-4H
SMILES ClC(=O)c1ccccc1Br
IUPAC Name 2-bromobenzoyl chloride
Molecular Weight (g/mol) 219.46
Formula Weight 217.9134
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.