missing translation for 'onlineSavingsMsg'
Learn More

2-Bromodibenzothiophene, 98%

Product Code. 15471929
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471929 5 g 5g
15461929 1 g 1g
15481929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471929 Supplier Thermo Scientific Alfa Aesar Supplier No. H64645.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22439-61-8 | C12H7BrS | 263.152 g/mol

Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22439-61-8
Molecular Formula C12H7BrS
Molecular Weight (g/mol) 263.152
MDL Number MFCD00089285
InChI Key IJICRIUYZZESMW-UHFFFAOYSA-N
Synonym 2-bromodibenzo b,d thiophene, dibenzothiophene, 2-bromo, 2-bromobenzo b benzo b thiophene, 4-bromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, acmc-209fwi, 2-bromo-dibenzothiophene, dibenzothiophene,2-bromo, 2-bromodibenzo b thiophene, 2-bromo-dibenzo b,d thiophene, dibenzo b,d thiophene, 2-bromo
PubChem CID 299508
IUPAC Name 2-bromodibenzothiophene
SMILES C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br

Specifications

Melting Point 123°C to 129°C (literature)
Quantity 5 g
Beilstein 142203
Solubility Information Slightly soluble in water.
Formula Weight 263.15
Percent Purity 98%
Chemical Name or Material 2-Bromodibenzothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.