missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethoxy)-tert-butyldimethylsilane, 98+%, stabilized, Thermo Scientific Chemicals

Product Code. 10122874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122874 1 g Glass bottle 1g
10784375 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10122874 Supplier Thermo Scientific Acros Supplier No. 399830010
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86864-60-0
Molecular Formula C8H19BrOSi
Molecular Weight (g/mol) 239.22
MDL Number MFCD00209550
InChI Key JBKINHFZTVLNEM-UHFFFAOYSA-N
Synonym 2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
PubChem CID 3608067
IUPAC Name 2-bromoethoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCBr

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color White to Yellow
Density 1.1150g/mL
Boiling Point 70°C to 75°C (2.5 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)CSi(CH3)2O(CH2)2Br
Quantity 1 g
Specific Gravity 1.115
Stabilizer 5 % max. Na2CO3
Formula Weight 239.22
Percent Purity 98+%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material (2-Bromoethoxy)-tert-butyldimethylsilane
Show More Show Less
compliance-icons
Product Identifier
  • (2-Bromoethoxy)-tert-butyldimethylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.