missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)benzene, 98%

Product Code. 11414194
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414194 500 g 500g
11404194 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414194 Supplier Thermo Scientific Alfa Aesar Supplier No. A12528.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-63-9 | C8H9Br | 185.064 g/mol

(2-Bromoethyl)benzene is used in the preparation of phenelzine by reacting with hydrazine. It is also used as a starting material to prepare various beta-phenethyl derivatives, active pharmaceutical ingredients, and fragrances. It finds application as a flame retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromoethyl)benzene is used in the preparation of phenelzine by reacting with hydrazine. It is also used as a starting material to prepare various beta-phenethyl derivatives, active pharmaceutical ingredients, and fragrances. It finds application as a flame retardant.

Solubility
Miscible with ether and benzene. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-63-9
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000240
InChI Key WMPPDTMATNBGJN-UHFFFAOYSA-N
Synonym 2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
PubChem CID 7666
IUPAC Name 2-bromoethylbenzene
SMILES C1=CC=C(C=C1)CCBr

Specifications

Melting Point -56°C
Density 1.355
Boiling Point 220°C to 221°C
Flash Point 89°C (192°F)
Linear Formula C6H5(CH2)2Br
Refractive Index 1.556
Quantity 500 g
Beilstein 507487
Solubility Information Miscible with ether and benzene. Immiscible with water.
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material (2-Bromoethyl)benzene
Show More Show Less
compliance-icons
Product Identifier
  • (2-Bromoethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: (2-Bromoethyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.