missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl ether, 90%

Product Code. 12188721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12188721 5 mL 5mL
12178721 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12188721 Supplier Thermo Scientific Acros Supplier No. 445860050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5414-19-7 | C4H8Br2O | 231.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5414-19-7
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.92
InChI Key FOZVXADQAHVUSV-UHFFFAOYSA-N
Synonym bis 2-bromoethyl ether, 1-bromo-2-2-bromoethoxy ethane, 2-bromoethyl ether, 2,2'-dibromodiethyl ether, ethane, 1,1'-oxybis 2-bromo, ether, bis 2-bromoethyl, beta,beta'-dibromodiethyl ether, 2,2-dibromodiethyl ether, 2-bromoethylether
PubChem CID 21521
IUPAC Name 1-bromo-2-(2-bromoethoxy)ethane
SMILES C(CBr)OCCBr

Specifications

Density 1.8450g/mL
Boiling Point 92°C to 93°C (12.0 mmHg)
Flash Point 85°C
Assay Percent Range 88% min. (GC)
Packaging Glass Bottle
Linear Formula [Br(CH2)2]2O
Quantity 5 mL
Specific Gravity 1.845
Formula Weight 231.92
Percent Purity 90%
Chemical Name or Material 2-Bromoethyl ether
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 2-Bromoethyl ether
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.