missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl phenyl sulfide, 97%

Product Code. 11333136
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333136 5 g 5g
11343136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333136 Supplier Thermo Scientific Alfa Aesar Supplier No. L11005.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4837-01-8 | C8H9BrS | 217.12 g/mol

2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4837-01-8
MDL Number MFCD00040826

Specifications

Density 1.44
Boiling Point 140°C to 141°C (10 mmHg)
Molecular Formula C12H18BNO2
Linear Formula C6H5SCH2CH2Br
Quantity 5 g
Beilstein 2206188
Synonym 2-2-phenylethyl-1,3,6,2-dioxazaborocane, 2-phenylethyl-1-boronic acid diethanolamine ester, 2-phenylethyl-1-boronic acid diethanolamine cyclic ester
Solubility Information Insoluble in water.
InChI Key NYYNBCZSIYCXIZ-UHFFFAOYSA-N
SMILES B1(OCCNCCO1)CCC2=CC=CC=C2
IUPAC Name 2-(2-phenylethyl)-1,3,6,2-dioxazaborocane
Molecular Weight (g/mol) 219.09
PubChem CID 24208882
Formula Weight 217.12
Percent Purity 97%
Chemical Name or Material 2-Bromoethyl phenyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.