missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)trimethylammonium Bromide 98%, Thermo Scientific Chemicals

Product Code. 10773393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10773393 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10773393 Supplier Thermo Scientific Acros Supplier No. 152350050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2758-06-7
Molecular Formula C5H13Br2N
Molecular Weight (g/mol) 246.97
MDL Number MFCD00011865
InChI Key OINMNSFDYTYXEQ-UHFFFAOYSA-M

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 240°C to 244°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula BrCH2CH2N(CH3)3Br
Quantity 5 g
Synonym 2-bromoethyl trimethylammonium bromide, 2-bromo-n,n,n-trimethylethanaminium bromide, 2-bromoethyltrimethylammonium bromide, bromcholin, hypercyl, kathesin, btab, btab the plant regulator, bcb the plant regulator, unii-kh1i7ru18l
SMILES C[N+](C)(C)CCBr.[Br-]
IUPAC Name 2-bromoethyl(trimethyl)azanium;bromide
PubChem CID 17689
Formula Weight 246.97
Percent Purity ≥97.5%
Physical Form Chunks and/or Powder
Chemical Name or Material (2-Bromoethyl)trimethylammonium bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (2-Bromoethyl)trimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.