missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethylamine hydrobromide, 99%

Product Code. 10476900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2.5kg
250g
25g
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10476900 25 g Plastic bottle 25g
10486900 100 g Plastic bottle 100g
10397143 250 g Plastic bottle 250g
10359043 500 g Plastic bottle 500g
10697513 1 kg Plastic bottle 1kg
11905171 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10476900 Supplier Thermo Scientific Acros Supplier No. 106930250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2576-47-8 | C2H6BrN·HBr | 204.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2576-47-8 , 107-09-5
Molecular Formula C2H6BrN·HBr
Molecular Weight (g/mol) 204.9
MDL Number MFCD00012886
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula BrCH2CH2NH2·HBr
Quantity 25 g
Beilstein 04,134
Solubility Information Solubility in water: >500g/L (20°C)
Formula Weight 204.9
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromoethylamine hydrobromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromoethylamine hydrobromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromoethylamine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.