missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoimidazole, 95%

Product Code. 15499287
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499287 1 g 1g
15409297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499287 Supplier Thermo Scientific Alfa Aesar Supplier No. H52553.03
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16681-56-4 | C3H3BrN2 | 146.98 g/mol

2-Bromoimidazole is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoimidazole is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16681-56-4
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD02179526
InChI Key AXHRGVJWDJDYPO-UHFFFAOYSA-N
Synonym 2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole
PubChem CID 2773261
IUPAC Name 2-bromo-1H-imidazole
SMILES BrC1=NC=CN1

Specifications

Melting Point 197°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 146.98
Percent Purity 95%
Chemical Name or Material 2-Bromoimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.