missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-1,4-benzodioxane, 97%

Product Code. 11339759
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11339759 25 g 25g
11319759 1 g 1g
11329759 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11339759

Brand: Thermo Scientific Alfa Aesar B22149.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2164-34-3 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2164-34-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00044831
InChI Key QYLFKNVZIFTCIY-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,4-benzodioxane, 2-bromomethyl-2,3-dihydro-1,4-benzodioxine, 2-bromomethyl-2,3-dihydrobenzo b 1,4 dioxine, 2-bromomethyl-2,3-dihydro-benzo 1,4 dioxine, 2-bromomethyl-2,3-dihydrobenzo 1,4 dioxine, 2-bromomethyl-2,3-dihydro-1,4-benzodioxin, 3-bromomethyl-2,3-dihydro-1,4-benzodioxine, acmc-1cuiw, 2-bromomethylbenzo-1,4-dioxan
PubChem CID 98333
IUPAC Name 3-(bromomethyl)-2,3-dihydro-1,4-benzodioxine
SMILES C1C(OC2=CC=CC=C2O1)CBr

Specifications

Boiling Point 143°C to 146°C (8 mmHg)
Refractive Index 1.5745
Quantity 25 g
Beilstein 150154
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-1,4-benzodioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.