missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine, 97%, Thermo Scientific™

Product Code. 11340607
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11340607 250 mg Amber Glass Bottle 250mg
10376804 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340607 Supplier Thermo Scientific Maybridge Supplier No. CC38008CB
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 869901-04-2
Molecular Formula C10H13BrN2
Molecular Weight (g/mol) 241.132
MDL Number MFCD09817465
InChI Key UFXKRINLXMWRES-UHFFFAOYSA-N
Synonym 2-bromomethyl-6-pyrrolidin-1-yl pyridine, 2-bromomethyl-6-pyrrolidin-1-ylpyridine, pyridine,2-bromomethyl-6-1-pyrrolidinyl, 6-bromomethyl-2-pyrrolidinylpyridine, 1-6-bromomethyl pyridin-2-yl pyrrolidine, pyridine, 2-bromomethyl-6-1-pyrrolidinyl
PubChem CID 24229482
IUPAC Name 2-(bromomethyl)-6-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=CC(=N2)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 241.13
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.