missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)benzonitrile, TRC

Product Code. 30340768
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340768 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340768 Supplier Toronto Research Chemicals Supplier No. TRCB68797010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Bromomethyl)benzonitrile
CAS 22115-41-9
Molecular Formula C8H6BrN
Quantity 10 g
Synonym alpha-Bromo-o-tolunitrile, (2-Cyanophenyl)methyl Bromide, 1-(Bromomethyl)-2-cyanobenzene, 2-(Bromomethyl)benzonitrile, 2-Cyano-a-bromotoluene, 2-Cyanobenzyl Bromide, o-(Bromomethyl)benzonitrile, o-Cyanobenzyl bromide, alpha-Bromo-2-cyanotoluene, alpha-Bromo-otolunitrile, alpha-Bromo-o-toluonitrile
InChI Formula InChI=1S/C8H6BrN/c9-5-7-3-1-2-4-8(7)6-10/h1-4H,5H2
SMILES BrCc1ccccc1C#N
IUPAC Name 2-(bromomethyl)benzonitrile
Molecular Weight (g/mol) 196.04
Formula Weight 194.9684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.