missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)pyridine hydrobromide, 98%

Product Code. 10581892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10581892 5 g Glass bottle 5g
10794753 25 g Glass bottle 25g
10606884 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10581892 Supplier Thermo Scientific Acros Supplier No. 370110050
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 31106-82-8 | C6H6BrN·HBr | 252.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31106-82-8
Molecular Formula C6H6BrN·HBr
Molecular Weight (g/mol) 252.94
MDL Number MFCD01863544
InChI Key JQDNCGRNPYKRAO-UHFFFAOYSA-N
Synonym 2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide
PubChem CID 2734720
IUPAC Name 2-(bromomethyl)pyridine;hydrobromide
SMILES C1=CC=NC(=C1)CBr.Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 149.0°C to 152.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrCH2(C5H3N)
Quantity 5 g
Formula Weight 252.94
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-(Bromomethyl)pyridine hydrobromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Bromomethyl)pyridine hydrobromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.