missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopalmitic Acid, TRC

Product Code. 30347525
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347525 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347525 Supplier Toronto Research Chemicals Supplier No. TRCB1826375G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-bromohexadecanoic acid
CAS 18263-25-7
Molecular Formula C16 H31 Br O2
Quantity 5 g
Synonym 2-Bromopalmitic Acid, 2-Bromohexadecanoic Acid, DL-2-Bromopalmitate, NSC 58378, alpha-Bromopalmitic Acid, 1-Bromopentadecanecarboxylic Acid, 2-Bromo-n-hexadecanoic Acid
InChI Formula InChI=1S/C16H31BrO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(17)16(18)19/h15H,2-14H2,1H3,(H,18,19)
SMILES CCCCCCCCCCCCCCC(Br)C(=O)O
IUPAC Name 2-bromohexadecanoic acid
Molecular Weight (g/mol) 335.32
Formula Weight 334.151
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.