missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Bromopropionic-2,3,3,3-d4 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Bromopropanoic Acid-D4 |
| CAS | 60153-93-7 |
| Molecular Formula | C3 D4 H Br O2 |
| Quantity | 0.5 g |
| Synonym | Propanoic-2,3,3,3-d4 acid, 2-bromo- (9 CI), 2-Bromopropanoic-2,3,3,3-d4 acid, 2-Bromopropanoic acid-2,3,3,3-d4, 2-Bromopropanoic Acid-D4, (±)-2-Bromopropionic-2,3,3,3-d4 Acid |
| InChI Formula | InChI=1 S/C3H5BrO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/i1D3,2 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Br)C(=O)O |
| IUPAC Name | 2-bromo-2,3,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 156.9993 |
| Formula Weight | 155.9724 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction