missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrazine, 95%

Product Code. 15492222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492222 1 g 1g
15402232 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492222 Supplier Thermo Scientific Acros Supplier No. 458160010
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 56423-63-3 | C4H3BrN2 | 159.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56423-63-3
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 159.99
InChI Key WGFCNCNTGOFBBF-UHFFFAOYSA-N
Synonym pyrazine, bromo, bromopyrazine, pyrazine, 2-bromo, 5-bromopyrazine, brom-pyrazin, 2-bromanylpyrazine, pubchem18036, acmc-209lsg, 2-bromopyrazine, pyrazine, bromo-9ci
PubChem CID 642800
IUPAC Name 2-bromopyrazine
SMILES C1=CN=C(C=N1)Br

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 159.99
Percent Purity 95%
Chemical Name or Material 2-Bromopyrazine
compliance-icons compliance-icons
Product Identifier
  • 2-Bromopyrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.