missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine, 99%

Product Code. 10793714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
Product Code. Quantity Packaging unitSize
10793714 500 mL Glass bottle 500mL
10674752 25 mL Glass bottle 25mL
10366070 100 mL Glass bottle 100mL
3 options
This item is not returnable. View return policy

Product Code. 10793714

Brand: Thermo Scientific Acros 107405000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-04-6 | C5H4BrN | 158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-04-6
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 158
MDL Number MFCD00006219
InChI Key IMRWILPUOVGIMU-UHFFFAOYSA-N
Synonym o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8
PubChem CID 7973
ChEBI CHEBI:51574
IUPAC Name 2-bromopyridine
SMILES C1=CC=NC(=C1)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 1.6500g/mL
Boiling Point 192°C to 194°C (761.0 mmHg)
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)Br
Refractive Index 1.5705 to 1.5725
Quantity 500 mL
Beilstein 20, 233
Specific Gravity 1.65
Solubility Information Solubility in water: 2.5g/L water (20°C)
Formula Weight 158
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromopyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.