missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotetradecane, 95%

Product Code. 11383106
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383106 5 g 5g
11393106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383106 Supplier Thermo Scientific Alfa Aesar Supplier No. L10974.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74036-95-6 | C14H29Br | 277.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74036-95-6
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.29
MDL Number MFCD00045033
InChI Key WZGQIDWFBFDMLE-UHFFFAOYSA-N
Synonym tetradecane, 2-bromo
PubChem CID 12798926
IUPAC Name 2-bromotetradecane
SMILES CCCCCCCCCCCCC(C)Br

Specifications

Boiling Point 160°C to 161°C (8 mmHg)
Refractive Index 1.46
Quantity 5 g
Beilstein 2071530
Formula Weight 277.3
Percent Purity 95%
Chemical Name or Material 2-Bromotetradecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.