missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene-3-carbonitrile, 97%, Thermo Scientific™

Product Code. 10467493
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10467493 250 mg Amber Glass Bottle 250mg
10337404 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10467493 Supplier Thermo Scientific Maybridge Supplier No. CC56616CB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 56182-43-5
Molecular Formula C5H2BrNS
Molecular Weight (g/mol) 188.042
MDL Number MFCD09064981
InChI Key GHMDOVNPFWGFSZ-UHFFFAOYSA-N
Synonym 2-bromo-3-cyanothiophene, 2-bromo-3-cyanothiophen, 2-bromo-3-thiophenecarbonitrile, 2-bromo-thiophene-3-carbonitrile, 3-thiophenecarbonitrile,2-bromo, 2-bromanylthiophene-3-carbonitrile, 3-thiophenecarbonitrile, 2-bromo
PubChem CID 12079026
IUPAC Name 2-bromothiophene-3-carbonitrile
SMILES C1=CSC(=C1C#N)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.05
Percent Purity 97%
Chemical Name or Material 2-Bromothiophene-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.