missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol, anhydrous, 99%, packaged under Argon in resealable ChemSeal bottles

Product Code. 11394847
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394847 1 L 1 L
11304857 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394847 Supplier Thermo Scientific Alfa Aesar Supplier No. 041871.K2
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-92-2 | C4H10O | 74.12 g/mol

2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point °C to 115°C
Color Colorless
Appearance Colorless liquid
Density 0.808 g/cm3 at 20°C
Quantity 1 L
Flash Point 26°C (79°F)
Ignition Point 390°C (734°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.397
Assay Percent Range 99%
Vapor Pressure 17.3 hPa (13mm Hg) at 20°C
Beilstein 1718765
Boiling Point 98°C to 101°C
Solubility Information Miscible with methanol and diethyl ether. Slightly miscible with water.
Formula Weight 74.12
Chemical Name or Material 2-Butanol, H2O <0.01%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.