missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol, Solstice

Product Code. 15612510
Change view
Click to view available options
Quantity:
30 kg
Packaging:
Stainless Steel Can
Unit Size:
30kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15612510 30 kg Stainless Steel Can 30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15612510 Supplier Fluka Supplier No. 2412630KG
  • ONLINE EXCLUSIVE PRICE
    589.00 €
    484.05 € / 30kg
    Save 104.95 €
    17% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 78-92-2
CAS Max % 100.0000%
Density 0.81 g/cm3
Quantity 30 kg
Flash Point 24°C
Packaging Stainless Steel Can
Molecular Formula C4H10O
MDL Number MFCD00004569
UN Number UN1120
Beilstein 773649
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
Boiling Point 99°C
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILES CCC(C)O
IUPAC Name butan-2-ol
Molecular Weight (g/mol) 74.12
PubChem CID 6568
ChEBI CHEBI:35687
Formula Weight 74.12
Percent Purity >99%
Grade Performance
Chemical Name or Material 2-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.