missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol, Reagent Grade, ≥99%, Honeywell™

Product Code. 15667700 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15667700 6 x 1 L Glass Bottle Pack of 6
15637700 1 L Glass Bottle 1 L
15647700 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15667700 Leverandør Honeywell Research Chemicals Leverandørnr. B859196X1L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Tekniske data

pH neutral
Melting Point -115°C
Color Colorless
Density 0.808g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.098
Flash Point 27°C (80.6°F)
Ignition Point 761°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3CH2CH(OH)CH3
Vapor Pressure 12.5mmHg (20°C)
UN Number UN1120
Beilstein 773649
Boiling Point 98°C
Formula Weight 74.12g/mol
Percent Purity ≥99%
Chemical Name or Material 2-Butanol
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • 2-Butanol
Signalord
  • Warning
Farekategori
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.