missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, 99+%, for electronic use (MOS), residue free

Product Code. 10113341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113341 1 L Glass Bottle 1 L
10687934 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113341 Supplier Thermo Scientific Acros Supplier No. 327910010
  • ONLINE EXCLUSIVE PRICE
    52.75 €
    51.20 € / 1 L
    Save 1.55 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78-93-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.8060g/mL
Quantity 1 L
Flash Point −7°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Residue after Evaporation 0.0005% max.
Assay Percent Range 99+%
Specific Gravity 0.806
Linear Formula C2H5COCH3
Viscosity 0.42 mPa.s (15°C)
Water 0.2% max. (Coulometric)
Beilstein 01, IV, 3243
Fieser 01,678
Merck Index 15, 6143
Boiling Point 80.0°C
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity ≥99%
Grade Electronic
Chemical Name or Material 2-Butanone, Residue Free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.