missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, 99+%, extra pure

Product Code. 10502551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10502551 25 mL Glass Bottle 25mL
10306510 1 L Glass Bottle 1 L
10367320 2.5 L Glass Bottle 2.5 L
10227333 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10502551 Supplier Thermo Scientific Acros Supplier No. 149670250
  • ONLINE EXCLUSIVE PRICE
    31.75 €
    30.80 € / 25mL
    Save 0.95 €
    2% Off
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 78-93-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Spezifikation

Melting Point -87.0°C
CAS Min % 99.0
Color Colorless
CAS Max % 100.0
Density 0.8060g/mL
Quantity 25 mL
Flash Point −7°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Residue after Evaporation 0.005% max.
Assay Percent Range 99+%
Specific Gravity 0.806
Linear Formula C2H5COCH3
Vapor Pressure Vapor Pressure: 105mbar at 20°C
Viscosity 0.42 mPa.s (15°C)
Water 0.2% max.
Beilstein 01, IV, 3243
Fieser 01,678
Merck Index 15, 6143
Boiling Point 80.0°C
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material 2-Butanone
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • 2-Butanone
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.