missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ACS Reagent, ≥99%, Solstice

Product Code. 15625070
Change view
Click to view available options
Quantity:
500 mL
2 L
2.5 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
2 L
2.5 L
20 L
500mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15625070 6 x 1 L Glass Bottle Pack of 6
15615070 500 mL Glass Bottle 500mL
15605070 2 L Glass Bottle 2 L
15685060 2.5 L Glass Bottle 2.5 L
15695060 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15625070 Supplier Honeywell-Riedel-de Haen Supplier No. 3604736X1L
  • ONLINE EXCLUSIVE PRICE
    468.00 €
    345.50 € / Pack of 6
    Save 122.50 €
    26% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -87°C
Density 0.805g/mL at 25°C
Quantity 6 x 1 L
Flash Point −3°C (26.6°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index n20/D 1.379
Boiling Point 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.