missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
100 mL
1 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
Pack of 4
Pack of 6
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -87°C (literature) |
| CAS Max % | 100.0000% |
| Density | 0.805 g/mL at 25 °C(lit.) |
| Boiling Point | 79°C to 81°C |
| Quantity | 4 x 2.5 L |
| Flash Point | −4°C |
| Packaging | Glass Bottle |
| Linear Formula | C2H5COCH3 |
| UN Number | UN1193 |
| Beilstein | 741880 |
| Show More |
Safety and Handling
Product Identifier
- 2-Butanone
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : butanone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction