missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyacetic acid, 98%

Product Code. 10359480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10359480 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10359480 Supplier Thermo Scientific Acros Supplier No. 257760010
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2516-93-0 | C6H12O3 | 132.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2516-93-0
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 108.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12O3
Refractive Index 1.4250 to 1.4270
MDL Number MFCD00067044
Quantity 1 g
Specific Gravity 1.01
Synonym butoxyacetic acid, n-butoxyacetic acid, acetic acid, butoxy, ccris 7195, acetic acid, 2-butoxy, 2-n-butoxy acetic acid, butoxyaceticacid, 3-oxaheptanoic acid, acmc-1cb1h, ajqoasgwdcbkcj-uhfffaoysa
InChI Key AJQOASGWDCBKCJ-UHFFFAOYSA-N
SMILES CCCCOCC(=O)O
IUPAC Name 2-butoxyacetic acid
Molecular Weight (g/mol) 132.15
PubChem CID 41958
Formula Weight 132.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butoxyacetic acid, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Butoxyacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.