missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyacetic Acid, TRC

Product Code. 30135791
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135791 500 mg 500mg
30133441 100 mg 100mg
30004989 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135791 Supplier Toronto Research Chemicals Supplier No. TRCB700378500MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-93-0
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
Synonym Acetic acid, butoxy- (6CI,7CI,8CI,9CI), Butoxy acetic acid, Butyloxyacetic acid, NSC 10980
IUPAC Name 2-butoxyacetic acid
SMILES CCCCOCC(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H12O3/c1-2-3-4-9-5-6(7)8/h2-5H2,1H3,(H,7,8)
Formula Weight 132.0786
Chemical Name or Material 2-Butoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.