missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 10089571
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10089571 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10089571 Supplier Thermo Scientific Maybridge Supplier No. SB02043DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 79047-41-9
Molecular Formula C8H13ClN2O
Molecular Weight (g/mol) 188.655
MDL Number MFCD01934397
InChI Key DXSZKDOOHOBZMT-UHFFFAOYSA-N
Synonym 2-butyl-4-chloro-1h-imidazol-5-yl methanol, 2-butyl-5-chloro-1h-imidazol-4-yl methanol, 2-butyl-5-chloroimidazole-4-methanol, 2-butyl-4-chloro-5-hydroxymethyl imidazole, 2-butyl-4-chloro-1h-imidazole-5-methanol, 2-butyl-5-chloro-1h-imidazol-4-yl-methanol, 2-butyl-5-chloro-3h-imidazol-4-yl methanol, 2-butyl-4-chloro-5-hydroxyimidazole, losartan impurity 1, acmc-1bk6v
PubChem CID 2735662
IUPAC Name (2-butyl-4-chloro-1H-imidazol-5-yl)methanol
SMILES CCCCC1=NC(=C(N1)CO)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.66
Percent Purity 97%
Chemical Name or Material (2-Butyl-5-chloro-1H-imidazol-4-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.