missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-d3-4-chloro-1H-imidazole-5-carboxaldehyde, TRC

Product Code. 30345017
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345017 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30345017 Supplier Toronto Research Chemicals Supplier No. TRCB69063225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butyl-d3-4-chloro-1H-imidazole-5-carboxaldehyde
CAS 1189927-81-8
Molecular Formula C8H8D3ClN2O
Quantity 25 mg
Synonym 2-Butyl-d3-5-chloro-1H-imidazole-4-carboxaldehyde, 2-Butyl-d3-4-chloro-5-formylimidazole, 2-Butyl-d3-4-chloroimidazole-5-carboxaldehyde, 2-Butyl-d3-5-formyl-4-chloroimidazole
InChI Formula InChI=1S/C8H11ClN2O/c1-2-3-4-7-10-6(5-12)8(9)11-7/h5H,2-4H2,1H3,(H,10,11)/i1D3
SMILES [2H]C([2H])([2H])CCCc1nc(C=O)c(Cl)[nH]1
IUPAC Name 5-chloro-2-(4,4,4-trideuteriobutyl)-1H-imidazole-4-carbaldehyde
Molecular Weight (g/mol) 189.66
Formula Weight 189.0748
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.