missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne, 98%

Product Code. 11424914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424914 100 g 100g
11404914 5 g 5g
11414914 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424914 Supplier Thermo Scientific Alfa Aesar Supplier No. A13823.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 503-17-3 | C4H6 | 54.092 g/mol

2-Butyne is used to synthesize alkylated hydroquinones in the total synthesis of Vitamin E. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyne is used to synthesize alkylated hydroquinones in the total synthesis of Vitamin E. It is also used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Avoid extremes of temperature and direct sunlight. Heat-sensitive. Avoid contact with oxidizing materials. Incompatible with Halogens. Chlorinated solvents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503-17-3
Molecular Formula C4H6
Molecular Weight (g/mol) 54.092
MDL Number MFCD00009275
InChI Key XNMQEEKYCVKGBD-UHFFFAOYSA-N
Synonym 2-butyne, dimethylacetylene, crotonylene, unii-lke6d3018e, ch3cequivcch3, 1,2-dimethylvinylene, 1-methyl-1-propynyl, 2-butyn, acmc-209kl9
PubChem CID 10419
IUPAC Name but-2-yne
SMILES CC#CC

Specifications

Melting Point -32°C
Density 0.69
Boiling Point 24°C to 27°C
Flash Point −49°C (−56°F)
Odor Recognizable
Refractive Index 1.392
Quantity 100 g
UN Number UN3295
Beilstein 1361340
Solubility Information Not miscible in water.
Formula Weight 54.09
Percent Purity 98%
Chemical Name or Material 2-Butyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.