missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxybenzaldehyde, 99%

Produktkod. 10107213 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
25 g
100 g
Packaging:
Glass bottle
Förpackningsstorlek:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging Förpackningsstorlek
10107213 25 g Glass bottle 25g
10236150 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107213 Supplier Thermo Scientific Acros Supplier No. 152080250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119-67-5 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-67-5
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00003336
InChI Key DYNFCHNNOHNJFG-UHFFFAOYSA-N
Synonym 2-carboxybenzaldehyde, phthalaldehydic acid, o-formylbenzoic acid, benzoic acid, 2-formyl, o-carboxybenzaldehyde, benzoic acid, formyl, benzaldehyde-2-carboxylic acid, formylbenzoic acid, carboxybenzaldehyde, pubchem7365
PubChem CID 8406
ChEBI CHEBI:17605
IUPAC Name 2-formylbenzoic acid
SMILES C1=CC=C(C(=C1)C=O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 96°C to 99°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HO2CC6H4CHO
Quantity 25 g
Beilstein 10, 666
Solubility Information Solubility in water: soluble
Formula Weight 150.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Carboxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Carboxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.