missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-1-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-1-ethanone, 97%, Thermo Scientific™

Product Code. 10788253
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10788253 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10788253 Supplier Thermo Scientific Maybridge Supplier No. MO07028DA
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 304685-89-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H13ClFNO
MDL Number MFCD01910944
Quantity 1 g
Synonym 2-chloro-1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-1-ethanone, 2-chloro-1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl ethanone, 2-chloro-1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl ethanone, 2-chloro-1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2-chloro-1-1-4-fluoro-phenyl-2,5-dimethyl-1h-pyrrol-3-yl-ethanone, 2-chloro-1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl ethan-1-one, acmc-209hg5, ethanone,2-chloro-1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl
InChI Key QNHMSVXIWDSTEP-UHFFFAOYSA-N
SMILES CC1=CC(=C(N1C2=CC=C(C=C2)F)C)C(=O)CCl
IUPAC Name 2-chloro-1-[1-(4-fluorophenyl)-2,5-dimethylpyrrol-3-yl]ethanone
Molecular Weight (g/mol) 265.712
PubChem CID 1537098
Formula Weight 265.71
Percent Purity 97%
Chemical Name or Material 2-chloro-1-[1-(4-fluorophenyl)-2, 5-dimethyl-1H-pyrrol-3-yl]-1-ethanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.